missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminohippuric acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC153805000

 View more versions of this product

Catalog No. AC153805000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Aminohippuric acid
Plastic bottle
198.0°C to 202.0°C
61-78-9
100.0
C9H10N2O3
MFCD00007890
15, 439
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, benzene, aceton and most, organic solvents, pract. insoluble in ccl4, very soluble in dilute hydrochloric acid and alkal
C1=CC(=CC=C1C(=O)NCC(=O)O)N
194.19
CHEBI:104011
Crystalline Powder
Authentic
Gray to White
500g
98.5
99%
H2NC6H4CONHCH2CO2H
14, II, 258
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
HSMNQINEKMPTIC-UHFFFAOYSA-N
2-[(4-aminobenzoyl)amino]acetic acid
2148
194.19
99%

Chemical Identifiers

61-78-9
194.19
HSMNQINEKMPTIC-UHFFFAOYSA-N
2148
2-[(4-aminobenzoyl)amino]acetic acid
C9H10N2O3
MFCD00007890
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
CHEBI:104011
C1=CC(=CC=C1C(=O)NCC(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents