Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminohippuric acid, 97%

$78.20 - $203.42

Chemical Identifiers

Chemical Name or Material 4-Aminohippuric acid
CAS 61-78-9
Molecular Formula C9H10N2O3
Formula Weight 194.19
MDL Number MFCD00007890
Beilstein 1213676
Merck Index 14,442
Synonym N-(4-Aminobenzoyl)glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1378614

  SDS
alfa aesar™
A1378614
25g Each for $78.20

AAA1378622

  SDS
alfa aesar™
A1378622
100g Each for $203.42
Description & Specifications

Specifications

Formula Weight 194.19
Chemical Name or Material 4-Aminohippuric acid
TSCA TSCA
Assay 97%
Molecular Formula C9H10N2O3
Melting Point ≈200°C (dec.)
MDL Number MFCD00007890
CAS 61-78-9
Quantity 25g
Beilstein 1213676
Merck Index 14,442
Synonym N-(4-Aminobenzoyl)glycine

Applications
4-Aminohippuric Acid is used in the measurement of renal plasma flow as a diagnostic tool which may be applied towards kidney disorders. It is used as muscle relaxant (smooth). It is also used to help differentiate and characterized organic anion transporter (OAT) isoforms.

Solubility
Soluble in alcohol, chloroform, benzene, and acetone. Insoluble in water, ether, carbon, and tetrachloride.

Notes
Store at -20°C. Light sensitive. Air sensitive. Always store away from incompatible compounds such as oxidizing agents.