missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Aminocarbonylphenylboronic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444900050
Specifications
4-Aminocarbonylphenylboronic acid | |
5g | |
96% min. (HPLC) | |
4-aminocarbonyl benzeneboronic acid, 4-aminocarbonyl phenylboronic acid, 4-aminocarbonylphenyl boronic acid, 4-aminocarbonylphenylboronic acid, 4-carbamoylbenzeneboronic acid, 4-carbamoylphenyl boronic acid, 4-dihydroxyboranyl benzamide, benzamide-4-boronic acid, boronic acid, 4-aminocarbonyl phenyl, p-aminocarbonyl phenylboronic acid | |
B(C1=CC=C(C=C1)C(=O)N)(O)O | |
164.96 | |
164.96 |
242.0°C to 252.0°C | |
123088-59-5 | |
C7H8BNO3 | |
GNRHNKBJNUVWFZ-UHFFFAOYSA-N | |
(4-carbamoylphenyl)boronic acid | |
2737811 | |
97% |
Chemical Identifiers
123088-59-5 | |
164.96 | |
4-aminocarbonyl benzeneboronic acid, 4-aminocarbonyl phenylboronic acid, 4-aminocarbonylphenyl boronic acid, 4-aminocarbonylphenylboronic acid, 4-carbamoylbenzeneboronic acid, 4-carbamoylphenyl boronic acid, 4-dihydroxyboranyl benzamide, benzamide-4-boronic acid, boronic acid, 4-aminocarbonyl phenyl, p-aminocarbonyl phenylboronic acid | |
(4-carbamoylphenyl)boronic acid |
C7H8BNO3 | |
GNRHNKBJNUVWFZ-UHFFFAOYSA-N | |
2737811 | |
B(C1=CC=C(C=C1)C(=O)N)(O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning