Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminocarbonyl-3-nitrobenzoic acid, 98%

$76.05 - $934.34

Chemical Identifiers

Chemical Name or Material 4-Aminocarbonyl-3-nitrobenzoic acid
CAS 87594-59-0
Molecular Formula C8H6N2O5
Formula Weight 210.15
MDL Number MFCD04039778
Synonym 4-Carboxy-2-nitrobenzamide; 3-Nitroterephthalamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2517803

 
alfa aesar™
B2517803
1g Each for $76.05

AAB2517814

 
alfa aesar™
B2517814
25g Each for $934.34
Description & Specifications

Specifications

Formula Weight 210.15
Chemical Name or Material 4-Aminocarbonyl-3-nitrobenzoic acid
Quantity 1g
Assay 98%
Molecular Formula C8H6N2O5
MDL Number MFCD04039778
CAS 87594-59-0
Synonym 4-Carboxy-2-nitrobenzamide; 3-Nitroterephthalamic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.