Please login to your online account to display your discounted pricing

4-Aminobutyraldehyde diethyl acetal, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC103260010

 View more versions of this product

Catalog No. AC103260010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 161.24
Physical Form Liquid
Quantity 1g
Color Clear colorless
Density 0.9000g/mL
CAS 1624116
Refractive Index 1.4270 to 1.4290 (20°C, 589nm)
Packaging Glass bottle
Linear Formula H2N(CH2)3CH(OC2H5)2
Synonym 4, 4-Diethoxybutylamine
Infrared Spectrum Authentic
MDL Number MFCD00008227
Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
Molecular Formula C8H19NO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
Assay ≥94%
Molecular Formula C8H19NO2
Linear Formula H2N(CH2)3CH(OC2H5)2
Formula Weight 161.24
MDL Number MFCD00008227
Synonym 4, 4-Diethoxybutylamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents