Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminobutyraldehyde diethyl acetal, 95%

Chemical Identifiers

Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
CAS 6346-09-4
Molecular Formula H2N(CH2)3CH(OCH2CH3)2
Formula Weight 161.25g/mol
MDL Number MFCD00008227
UN Number UN2735
Beilstein 1739649
Synonym 4-Aminobutanal diethyl acetal; 1-Amino-4,4-diethoxybutane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1428606

 
alfa aesar™
L14286-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1428614

 
alfa aesar™
L14286-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 196° to 198°C
Quantity 5g
Flash Point 63°C (145°F)
Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
CAS 6346-09-4
Assay 95%
Molecular Formula H2N(CH2)3CH(OCH2CH3)2
Formula Weight 161.25g/mol
MDL Number MFCD00008227
UN Number UN2735
Beilstein 1739649
Synonym 4-Aminobutanal diethyl acetal; 1-Amino-4,4-diethoxybutane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Applications
4-Aminobutyraldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agent, air.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.