missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Aminobiphenyl, 98%

Manufacturer:  Alfa Aesar™ H5542403

 View more versions of this product

Catalog No. AAH5542403



Description

Description

Applications
It is used to manufacture azo dyes.

Solubility
Soluble in Dichloromethane, DMSO and Methanol

Notes
Store at -20°C. Avoid heat and oxidizing agents.
Specifications

Specifications

4-Aminobiphenyl
52°C to 54°C
92-67-1
MFCD00007879
386533
1,1'-biphenyl-4-amine
DMVOXQPQNTYEKQ-UHFFFAOYSA-N
4-phenylaniline
7102
169.23
191°C (15mmHg)
1g
C12H11N
UN3077
14,1235
Soluble in Dichloromethane,DMSO and Methanol
C1=CC=C(C=C1)C2=CC=C(C=C2)N
169.227
CHEBI:1784
98%

Chemical Identifiers

92-67-1
169.227
DMVOXQPQNTYEKQ-UHFFFAOYSA-N
7102
4-phenylaniline
C12H11N
MFCD00007879
1,1'-biphenyl-4-amine
CHEBI:1784
C1=CC=C(C=C1)C2=CC=C(C=C2)N
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-177-1

RTECSNumber : DU8925000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents