Please login to your online account to display your discounted pricing

4-Aminobenzylamine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391450050

 View more versions of this product

Catalog No. AC391450050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Oily Liquid
Formula Weight 122.17
Quantity 5g
Color Clear light brown to brown
Density 1.0780g/mL
CAS Max % 100.0
Linear Formula NH2CH2C6H4NH2
Refractive Index 1.6080 to 1.6110 (20°C, 589nm)
Chemical Name or Material 4-Aminobenzylamine
Assay Percent Range 98%
Molecular Formula C7H10N2
CAS 4403-71-8
CAS Min % 97.5
MDL Number MFCD00075513
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Aminobenzylamine
CAS 4403-71-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H10N2
Linear Formula NH2CH2C6H4NH2
Formula Weight 122.17
MDL Number MFCD00075513
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents