missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Aminobenzoyl)hydrazide 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 103201000

 View more versions of this product

Catalog No. AC103201000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
151.17
White or beige
224° to 228°C
100.0
4-Aminobenzoic acid hydrazide
95%
MFCD00007606
C7H9N3O
H2NC6H4CONHNH2
Crystals or Powder
100g
14, I, 570
94.0
Authentic
Plastic bottle
95%
(4-Aminobenzoyl)hydrazide
5351-17-7

Chemical Identifiers

5351-17-7
100.0
C7H9N3O
151.17
4-Aminobenzoic acid hydrazide
≥94%
94.0
95%
H2NC6H4CONHNH2
MFCD00007606
(4-Aminobenzoyl)hydrazide, 95%

Safety and Handling

SDS & Safety Documents

Warning

EINECSNumber : 226-324-4

RTECSNumber : DG2580000