missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4-Aminobenzoyl)hydrazide 95%, ACROS Organics™

Manufacturer:  Acros Organics 103200050

 View more versions of this product

Catalog No. AC103200050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(4-Aminobenzoyl)hydrazide
Glass Bottle
224.0°C to 228.0°C
95%
94.0
95%
H2NC6H4CONHNH2
14, I, 570
WPBZMCGPFHZRHJ-UHFFFAOYSA-N
4-aminobenzohydrazide
21450
Crystals or Powder
Authentic
Beige or White
5g
5351-17-7
100.0
C7H9N3O
MFCD00007606
4-aminobenzhydrazide
C1=CC(=CC=C1C(=O)NN)N
151.169
151.17
95%

Chemical Identifiers

5351-17-7
151.169
WPBZMCGPFHZRHJ-UHFFFAOYSA-N
21450
C1=CC(=CC=C1C(=O)NN)N
C7H9N3O
MFCD00007606
4-aminobenzhydrazide
4-aminobenzohydrazide
Safety and Handling

Safety and Handling

Warning

EINECSNumber : 226-324-4

RTECSNumber : DG2580000

SDS Documents