missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Aminobenzoyl)hydrazide, 95%, ACROS Organics™
Chemical Identifiers
CAS | 5351-17-7 |
---|---|
Molecular Formula | C7H9N3O |
Molecular Weight (g/mol) | 151.17 |
MDL Number | MFCD00007606 |
InChI Key | WPBZMCGPFHZRHJ-UHFFFAOYSA-N |
Synonym | 4-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, 4-aminobenzoylhydrazine, amben hydrazide, aminostimil, benzoic acid, 4-amino-, hydrazide, p-aminobenzhydrazide, p-aminobenzoic acid hydrazide, p-aminobenzoic hydrazide, p-aminobenzoyl hydrazide |
PubChem CID | 21450 |
IUPAC Name | 4-aminobenzohydrazide |
SMILES | C1=CC(=CC=C1C(=O)NN)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC103200050
|
Thermo Fisher Scientific
103200050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
5351-17-7 | |
151.17 | |
WPBZMCGPFHZRHJ-UHFFFAOYSA-N | |
21450 | |
C1=CC(=CC=C1C(=O)NN)N |
C7H9N3O | |
MFCD00007606 | |
4-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, 4-aminobenzoylhydrazine, amben hydrazide, aminostimil, benzoic acid, 4-amino-, hydrazide, p-aminobenzhydrazide, p-aminobenzoic acid hydrazide, p-aminobenzoic hydrazide, p-aminobenzoyl hydrazide | |
4-aminobenzohydrazide |
Specifications
Authentic | |
Beige or White | |
5g | |
94.0 | |
94% min. (HPLC) | |
H2NC6H4CONHNH2 | |
14, I, 570 | |
WPBZMCGPFHZRHJ-UHFFFAOYSA-N | |
4-aminobenzohydrazide | |
21450 | |
Crystals or Powder | |
(4-Aminobenzoyl)hydrazide, 95% |
Glass Bottle | |
224.0°C to 228.0°C | |
5351-17-7 | |
100.0 | |
C7H9N3O | |
MFCD00007606 | |
4-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, 4-aminobenzoylhydrazine, amben hydrazide, aminostimil, benzoic acid, 4-amino-, hydrazide, p-aminobenzhydrazide, p-aminobenzoic acid hydrazide, p-aminobenzoic hydrazide, p-aminobenzoyl hydrazide | |
C1=CC(=CC=C1C(=O)NN)N | |
151.17 | |
151.17 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 226-324-4
RTECSNumber : DG2580000