missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminobenzotrifluoride hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 174180250

 View more versions of this product

Catalog No. AC174180250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
197.59
Yellow
100.0
MFCD00007897
C7H6F3N·HCl
H2NC6H4CF3·HCl
90774-69-9
4-(Trifluoromethyl)aniline hydrochloride
Granular Powder
25g
96.0
Glass bottle
4-Aminobenzotrifluoride hydrochloride
97%
97%
Authentic

Chemical Identifiers

90774-69-9
100.0
C7H6F3N·HCl
197.59
4-(Trifluoromethyl)aniline hydrochloride
≥96.0%
96.0
97%
H2NC6H4CF3·HCl
MFCD00007897
4-Aminobenzotrifluoride hydrochloride, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 292-679-7