Please login to your online account to display your discounted pricing

4-Aminobenzotrifluoride hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminobenzotrifluoride hydrochloride
Name Note 97%
CAS 90774-69-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H6F3N·HCl
Linear Formula H2NC6H4CF3·HCl
Formula Weight 197.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC174180250

  SDS
thermo fisher scientific
174180250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC174180050

  SDS
thermo fisher scientific
174180050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Granular Powder
Formula Weight 197.59
Quantity 25g
Color Yellow
EINECS Number 292-679-7
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Aminobenzotrifluoride hydrochloride
Packaging Glass bottle
Molecular Formula C7H6F3N·HCl
MDL Number MFCD00007897
Name Note 97%
Assay Percent Range 97%
Linear Formula H2NC6H4CF3·HCl
CAS 90774-69-9
Synonym 4-(Trifluoromethyl)aniline hydrochloride
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic