Learn More
4-Aminobenzotrifluoride, 99%, Thermo Scientific Chemicals
$80.39 - $80.39
Chemical Identifiers
CAS | 455-14-1 |
---|---|
Molecular Formula | C7H6F3N |
Molecular Weight (g/mol) | 161.13 |
MDL Number | MFCD00064396 |
InChI Key | ODGIMMLDVSWADK-UHFFFAOYSA-N |
Synonym | 4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine |
PubChem CID | 9964 |
ChEBI | CHEBI:40750 |
IUPAC Name | 4-(trifluoromethyl)aniline |
SMILES | C1=CC(=CC=C1C(F)(F)F)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC208910100
|
Thermo Scientific Chemicals
208910100 |
10 mL | Glass bottle |
Each for $80.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
455-14-1 | |
161.13 | |
ODGIMMLDVSWADK-UHFFFAOYSA-N | |
9964 | |
4-(trifluoromethyl)aniline |
C7H6F3N | |
MFCD00064396 | |
4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine | |
CHEBI:40750 | |
C1=CC(=CC=C1C(F)(F)F)N |
Specifications
455-14-1 | |
100.0 | |
Colorless to Yellow | |
83.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4830 to 1.4850 | |
MFCD00064396 | |
12, III, 2151 | |
4-trifluoromethyl aniline, 4-aminobenzotrifluoride, p-aminobenzotrifluoride, p-trifluoromethylaniline, benzenamine, 4-trifluoromethyl, 4-trifluoromethyl benzenamine, p-trifluoromethyl aniline, aniline, p-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluidine, a,a,a-trifluoro-p-toluidine | |
ODGIMMLDVSWADK-UHFFFAOYSA-N | |
4-(trifluoromethyl)aniline | |
9964 | |
161.13 | |
Liquid |
98.5 | |
5.0°C to 8.0°C | |
1.2960g/mL | |
86°C | |
98.5% min. (GC) | |
C7H6F3N | |
H2NC6H4CF3 | |
10 mL | |
1.296 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ether,methanol,ethanol and,aromatics | |
C1=CC(=CC=C1C(F)(F)F)N | |
161.13 | |
CHEBI:40750 | |
99% | |
4-Aminobenzotrifluoride, 99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minu
GHS Signal Word: Danger
EINECSNumber : 207-236-5
RTECSNumber : XU9260000
TSCA : TSCA
RUO – Research Use Only