Please login to your online account to display your discounted pricing

4-Aminobenzophenone 98%, ACROS Organics™


Quantity 25g

Manufacturer: fisher scientific acros  103170250

 View more versions of this product

Catalog No. AC103170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 197.24
Physical Form Fine Crystalline Powder
Quantity 25g
Color Yellow
Beilstein 14, 81
TSCA TSCA
EINECS Number 214-506-6
Melting Point 120° to 124°C
RTECS Number DJ0175000
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C13H11NO
Assay Percent Range 98%
Linear Formula H2NC6H4COC6H5
CAS 1137-41-3
Name Note 98%
Chemical Name or Material 4-Aminobenzophenone
MDL Number MFCD00007895
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

Specifications

Chemical Name or Material 4-Aminobenzophenone
Name Note 98%
CAS 1137-41-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H11NO
Linear Formula H2NC6H4COC6H5
Formula Weight 197.24
MDL Number MFCD00007895
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents