Learn More
4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1408906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Aminobenzamidine dihydrochloride | |
>300°C | |
MFCD00013001 | |
3692927 | |
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag | |
C1=CC(=CC=C1C(=N)N)N.Cl.Cl | |
208.086 | |
CHEBI:10618 | |
97% |
2498-50-2 | |
C7H11Cl2N3 | |
5 g | |
Hygroscopic | |
GHEHNICLPWTXJC-UHFFFAOYSA-N | |
4-aminobenzenecarboximidamide;dihydrochloride | |
75626 | |
208.09 |
Chemical Identifiers
2498-50-2 | |
208.086 | |
GHEHNICLPWTXJC-UHFFFAOYSA-N | |
75626 | |
4-aminobenzenecarboximidamide;dihydrochloride |
C7H11Cl2N3 | |
MFCD00013001 | |
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag | |
CHEBI:10618 | |
C1=CC(=CC=C1C(=N)N)N.Cl.Cl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 219-692-2
RTECSNumber : CV6126500
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only