Please login to your online account to display your discounted pricing

4-Aminobenzamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Aminobenzamide
CAS 2835-68-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O
Linear Formula H2NC6H4CONH2
Formula Weight 136.15
MDL Number MFCD00007999
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351645000

  SDS
thermo fisher scientific
AC351645000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC351640250

  SDS
fisher scientific acros
351640250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC351641000

  SDS
thermo fisher scientific
351641000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 136.15
Quantity 500g
Color Off-white to beige
Melting Point 181° to 185°C
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula H2NC6H4CONH2
CAS 2835-68-9
Chemical Name or Material 4-Aminobenzamide
Molecular Formula C7H8N2O
Assay Percent Range 98%
MDL Number MFCD00007999
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.