missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Aminobenzamide, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 351640250

 View more versions of this product

Catalog No. AC351640250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Aminobenzamide
97.5
Authentic
Crystalline Powder
98%
Off-white to beige
H2NC6H4CONH2
25g
2835-68-9
100.0
Glass bottle
≥97.5%
181° to 185°C
C7H8N2O
136.15
MFCD00007999

Chemical Identifiers

2835-68-9
100.0
C7H8N2O
136.15
4-Aminobenzamide
97.5
98%
H2NC6H4CONH2
MFCD00007999
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.