Learn More
4-Aminobenzamide, 98+%, Thermo Scientific Chemicals
$51.37 - $518.25
Chemical Identifiers
CAS | 2835-68-9 |
---|---|
Molecular Formula | C7H8N2O |
Molecular Weight (g/mol) | 136.154 |
MDL Number | MFCD00007999 |
InChI Key | QIKYZXDTTPVVAC-UHFFFAOYSA-N |
Synonym | p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4 |
PubChem CID | 76079 |
IUPAC Name | 4-aminobenzamide |
SMILES | C1=CC(=CC=C1C(=O)N)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1259414
|
Thermo Scientific Chemicals
A1259414 |
25 g |
N/A
|
|
|||||
AAA1259422
|
Thermo Scientific Chemicals
A1259422 |
100 g |
N/A
|
|
|||||
AAA1259436
|
Thermo Scientific Chemicals
A1259436 |
500 g |
N/A
|
|
|||||
Description
4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
2835-68-9 | |
136.154 | |
QIKYZXDTTPVVAC-UHFFFAOYSA-N | |
76079 | |
C1=CC(=CC=C1C(=O)N)N |
C7H8N2O | |
MFCD00007999 | |
p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4 | |
4-aminobenzamide |
Specifications
2835-68-9 | |
C7H8N2O | |
25 g | |
p-aminobenzamide, benzamide, 4-amino, 4-amino-benzamide, p-carbamoylaniline, p-aminobenzoic acid amide, benzamide, p-amino, 4-amino benzamide, unii-77722i6pac, ccris 6792, aminobenzamide-4 | |
QIKYZXDTTPVVAC-UHFFFAOYSA-N | |
4-aminobenzamide | |
76079 | |
≥98% |
182°C to 184°C | |
MFCD00007999 | |
2079485 | |
Slightly soluble in water. | |
C1=CC(=CC=C1C(=O)N)N | |
136.154 | |
136.15 | |
4-Aminobenzamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 220-612-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only