Please login to your online account to display your discounted pricing

4-Aminoantipyrine, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$232.41 - $232.41

Chemical Identifiers

Chemical Name or Material 4-Aminoantipyrine
CAS 83-07-8
Molecular Formula C11H13N3O
Formula Weight 203.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002104

 
avantor performance
A63005
Wide Mouth Amber Glass Bottle 100g Each for $232.41
Description & Specifications

Specifications

Formula Weight 203.24
Chemical Name or Material 4-Aminoantipyrine
CAS 83-07-8
Quantity 100g
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C11H13N3O