Learn More
4-Aminoantipyrine, 98%, ACROS Organics™
4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow
Manufacturer: Thermo Fisher Scientific AC103150025
Description
4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow
Specifications
(5% aq. soln.) clear | |
0.8000g/mL | |
1.5% max. | |
0.8 | |
Brown-Yellow to Yellow | |
2.5kg | |
83-07-8 | |
100.0 | |
C11H13N3O | |
24,273 | |
4-aminoantipyrene, 4-aminoantipyrine, 4-aminophenazone, aminoantipyrin, aminoantipyrine, aminoazophene, ampyrone, metapirazone, solnapyrin-a, solvapyrin-a | |
RLFWWDJHLFCNIJ-UHFFFAOYSA-N | |
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one | |
2151 | |
203.25 | |
98% |
4-Aminoantipyrine | |
Authentic | |
Plastic Bottle | |
0.1% max. | |
105.5°C to 110.0°C | |
98% | |
97.5 | |
98% | |
MFCD00003145 | |
15,413 | |
(5% in water) Clear | |
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N | |
203.25 | |
CHEBI:59026 | |
Powder or Crystals |
Chemical Identifiers
83-07-8 | |
203.25 | |
RLFWWDJHLFCNIJ-UHFFFAOYSA-N | |
2151 | |
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one |
C11H13N3O | |
MFCD00003145 | |
4-aminoantipyrene, 4-aminoantipyrine, 4-aminophenazone, aminoantipyrin, aminoantipyrine, aminoazophene, ampyrone, metapirazone, solnapyrin-a, solvapyrin-a | |
CHEBI:59026 | |
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 201-452-3
RTECSNumber : CD2480000
TSCA : TSCA