missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Aminoacetophenone, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC103095000

 View more versions of this product

Catalog No. AC103095000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
≥98.5%
Slightly yellow to brown
14, 46
04,18
Authentic
4'-Aminoacetophenone
99-92-3
99%
MFCD00007896
135.17
500g
104° to 107°C
14, 413
100.0
H2NC6H4COCH3
C8H9NO
98.5
Glass bottle

Chemical Identifiers

99-92-3
100.0
C8H9NO
135.17
4'-Aminoacetophenone
98.5
99%
H2NC6H4COCH3
MFCD00007896
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.