Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Aminoacetanilide, 98+%

$109.54 - $417.38

Chemical Identifiers

Chemical Name or Material 4'-Aminoacetanilide
CAS 122-80-5
Molecular Formula C8H10N2O
Formula Weight 150.18
MDL Number MFCD00007853
Beilstein 742888
Merck Index 14,411
Synonym 4-Acetamidoaniline; N-Acetyl-1,4-phenylenediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2074036

  SDS
alfa aesar™
B2074036
500g Each for $417.38

AAB2074022

  SDS
alfa aesar™
B2074022
100g Each for $109.54
Description & Specifications

Specifications

Formula Weight 150.18
Chemical Name or Material 4'-Aminoacetanilide
TSCA TSCA
Assay ≥98%
Molecular Formula C8H10N2O
MDL Number MFCD00007853
CAS 122-80-5
Beilstein 742888
Melting Point 161° to 164°C
Merck Index 14,411
Synonym 4-Acetamidoaniline; N-Acetyl-1,4-phenylenediamine
Boiling Point 267°C
Quantity 500g
Flash Point 195°C (383°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AD8225000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.