missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Aminoacetanilide, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 400601000

Catalog No. AC400601000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4'-Aminoacetanilide
122-80-5
100.0
>150°C
95%
C8H10N2O
MFCD00007853
15, 405
Solubility in water: soluble in cold water. Other solubilities: freely soluble in hot water,freely soluble in alcohol,ether
CC(=O)NC1=CC=C(C=C1)N
150.18
150.18
95%
94.0
267.0°C
Authentic
Glass bottle
NHCOCH3C6H4NH2
100g
4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
CHMBIJAOCISYEW-UHFFFAOYSA-N
N-(4-aminophenyl)acetamide
31230
95%
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persi

GHS Signal Word: Danger

EINECSNumber : 204-576-6

RTECSNumber : AD8225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only