missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Aminoacetanilide, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 400600250

 View more versions of this product

Catalog No. AC400600250


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4'-Aminoacetanilide
122-80-5
100.0
Brown to Pink
>150°C
95%
C8H10N2O
MFCD00007853
15, 405
Solubility in water: soluble in cold water. Other solubilities: freely soluble in hot water,freely soluble in alcohol,ether
CC(=O)NC1=CC=C(C=C1)N
150.18
150.18
Fine Crystalline Powder
95%
94.0
159.0°C to 165.0°C
267.0°C
Authentic
Glass bottle
NHCOCH3C6H4NH2
25 g
4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
CHMBIJAOCISYEW-UHFFFAOYSA-N
N-(4-aminophenyl)acetamide
31230
95%

Chemical Identifiers

122-80-5
150.18
CHMBIJAOCISYEW-UHFFFAOYSA-N
31230
CC(=O)NC1=CC=C(C=C1)N
C8H10N2O
MFCD00007853
4'-aminoacetanilide, n-4-aminophenyl acetamide, p-aminoacetanilide, 4-aminoacetanilide, 4-acetamidoaniline, p-amino acetanilide, p-acetamidoaniline, acetparamin, acetamide, n-4-aminophenyl, fourrine a
N-(4-aminophenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persi

GHS Signal Word: Danger

EINECSNumber : 204-576-6

RTECSNumber : AD8225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only