Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-N-methyl-alpha-toluenesulfonamide, 97%

Chemical Identifiers

Chemical Name or Material 4-Amino-N-methyl-alpha-toluenesulfonamide
CAS 109903-35-7
Molecular Formula C8H12N2O2S
Formula Weight 200.26
MDL Number MFCD00671697
UN Number UN3077
Beilstein 5865321
Synonym 4-Amino-N-methylbenzylsulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1458903

  SDS
alfa aesar™
L1458903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.26
MDL Number MFCD00671697
Chemical Name or Material 4-Amino-N-methyl-alpha-toluenesulfonamide
CAS 109903-35-7
UN Number UN3077
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Molecular Formula C8H12N2O2S
Melting Point 140° to 142°C
Synonym 4-Amino-N-methylbenzylsulfonamide
Beilstein 5865321
Quantity 1g
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.