missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-N-Fmoc-L-phenylalanine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62505MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-N-Fmoc-L-phenylalanine | |
C24H22N2O4 | |
250 mg | |
VALNSJHHRPSUDO-QFIPXVFZSA-N | |
(2S)-3-(4-aminophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid | |
7019927 | |
98% |
95753-56-3 | |
MFCD00237664 | |
fmoc-4-amino-l-phenylalanine, pubchem12464, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, a-fmoc-4-amino-l-phenylalanine, fmoc-4-amino-phenylalanine, fmoc-4-amino-phenylalanine, l-phenylalanine, 4-amino-n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-4-ao-l-phenylalanine, fmoc-phe p-nh2-oh, fmoc-phe p-nh2-oh | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)N)C(=O)O | |
402.45 | |
402.44 |
Chemical Identifiers
95753-56-3 | |
402.45 | |
VALNSJHHRPSUDO-QFIPXVFZSA-N | |
7019927 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)N)C(=O)O |
C24H22N2O4 | |
MFCD00237664 | |
fmoc-4-amino-l-phenylalanine, pubchem12464, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, a-fmoc-4-amino-l-phenylalanine, fmoc-4-amino-phenylalanine, fmoc-4-amino-phenylalanine, l-phenylalanine, 4-amino-n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-4-ao-l-phenylalanine, fmoc-phe p-nh2-oh, fmoc-phe p-nh2-oh | |
(2S)-3-(4-aminophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only