Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-8-(trifluoromethyl)quinoline, 97%

$104.68 - $363.27

Chemical Identifiers

Chemical Name or Material 4-Amino-8-(trifluoromethyl)quinoline
CAS 243977-15-3
Molecular Formula C10H7F3N2
Formula Weight 212.18
MDL Number MFCD00269346
Synonym 8-Trifluoromethyl-4-quinolinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5019303

 
alfa aesar™
H5019303
1g Each for $363.27

AAH50193MD

 
alfa aesar™
H50193MD
250mg Each for $104.68
Description & Specifications

Specifications

Formula Weight 212.18
Chemical Name or Material 4-Amino-8-(trifluoromethyl)quinoline
Assay 97%
Molecular Formula C10H7F3N2
MDL Number MFCD00269346
CAS 243977-15-3
Quantity 1g
Synonym 8-Trifluoromethyl-4-quinolinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Amino-8-(trifluoromethyl)quinoline is used to synthesis of rubber chemical, antiseptic, antipyretic, and antiperiodic properties and are used as antimalarials and for preparing other antimalarial drugs

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.