Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-7-(trifluoromethyl)quinoline, 98%

$107.43 - $372.50

Chemical Identifiers

Chemical Name or Material 4-Amino-7-(trifluoromethyl)quinoline
CAS 243666-11-7
Molecular Formula C10H7F3N2
Formula Weight 212.18
MDL Number MFCD00269411
Synonym 7-Trifluoromethyl-4-quinolinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5035503

 
alfa aesar™
H5035503
1g Each for $372.50

AAH50355MD

 
alfa aesar™
H50355MD
250mg Each for $107.43
Description & Specifications

Specifications

Formula Weight 212.18
Chemical Name or Material 4-Amino-7-(trifluoromethyl)quinoline
Assay 98%
Molecular Formula C10H7F3N2
MDL Number MFCD00269411
CAS 243666-11-7
Quantity 1g
Synonym 7-Trifluoromethyl-4-quinolinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.