Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine, 94%

$32.78 - $84.81

Chemical Identifiers

Chemical Name or Material 4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine
CAS 23771-52-0
Molecular Formula C5H5N5S
Formula Weight 167.19
MDL Number MFCD00005690
Beilstein 152193
Synonym 4-Amino-1,7-dihydro-6H-pyrazolo[3,4-d]pyrimidine-6-thione; 4-Amino-1H-pyrazolo[3,4-d]pyrimidine-6-thiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0693403

 
alfa aesar™
L0693403
1g Each for $84.81

AAL06934MD

 
alfa aesar™
L06934MD
250mg Each for $32.78
Description & Specifications

Specifications

Formula Weight 167.19
Assay 94%
MDL Number MFCD00005690
Molecular Formula C5H5N5S
Melting Point >300°C
CAS 23771-52-0
Beilstein 152193
Synonym 4-Amino-1,7-dihydro-6H-pyrazolo[3,4-d]pyrimidine-6-thione; 4-Amino-1H-pyrazolo[3,4-d]pyrimidine-6-thiol
Chemical Name or Material 4-Amino-6-mercapto-1H-pyrazolo[3,4-d]pyrimidine
Quantity 1g