Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-6-hydroxypyrimidine, 98%

$90.05 - $771.18

Chemical Identifiers

Chemical Name or Material 4-Amino-6-hydroxypyrimidine
CAS 1193-22-2
Molecular Formula C4H5N3O
Formula Weight 111.1
MDL Number MFCD00051502
Beilstein 112429
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0382506

 
alfa aesar™
L0382506
5g Each for $771.18

AAL03825MD

 
alfa aesar™
L03825MD
250mg Each for $90.05
Description & Specifications

Specifications

Formula Weight 111.1
Chemical Name or Material 4-Amino-6-hydroxypyrimidine
CAS 1193-22-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C4H5N3O
Melting Point 262° to 264°C
MDL Number MFCD00051502
Quantity 5g
Beilstein 112429
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.