Please login to your online account to display your discounted pricing

4-Amino-6-hydroxy-2-mercaptopyrimidine Monohydrate 98%, Acros Organics


Quantity 50g

Manufacturer: fisher scientific acros  103820500

 View more versions of this product

Catalog No. AC103820500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Amorphous Powder
Formula Weight 161.18
Quantity 50g
Color White to very slightly yellow
Beilstein 24, 476
RTECS Number UW0495000
CAS Max % 100.0
Name Note 98%
Molecular Formula C4H5N3OS·H2O
CAS 65802-56-4
CAS Min % 97.5
Assay Percent Range 98%
Chemical Name or Material 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate
MDL Number MFCD00150556
Infrared Spectrum Authentic
Synonym 6-Amino-2-thiouracil monohydrate

Specifications

Chemical Name or Material 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate
Name Note 98%
CAS 65802-56-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H5N3OS·H2O
Formula Weight 161.18
MDL Number MFCD00150556
Synonym 6-Amino-2-thiouracil monohydrate