missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-6-hydroxy-2-mercaptopyrimidine Monohydrate 98%, Acros Organics

Chemical Identifiers

CAS 65802-56-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C4H5N3OS·H2O
Formula Weight 161.18
MDL Number MFCD00150556
Synonym 6-Amino-2-thiouracil monohydrate
Chemical Name or Material 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC103820500 SDS Fisher Scientific Acros
103820500
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

65802-56-4
100.0
C4H5N3OS·H2O
MFCD00150556
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98%
97.5
98%
161.18
6-Amino-2-thiouracil monohydrate
≥97.5%

Specifications

65802-56-4
100.0
Amorphous Powder
C4H5N3OS·H2O
MFCD00150556
50g
6-Amino-2-thiouracil monohydrate
≥97.5%
97.5
Authentic
98%
161.18
White to very slightly yellow
24, 476
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98%

Safety and Handling

RTECSNumber : UW0495000