missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98+%, Thermo Scientific Chemicals
$129.90 - $129.90
Chemical Identifiers
CAS | 65802-56-4 |
---|---|
Molecular Formula | C4H7N3O2S |
Molecular Weight (g/mol) | 161.179 |
MDL Number | MFCD00150556 |
InChI Key | BWYGMIYEADAIGW-UHFFFAOYSA-N |
Synonym | 4-amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 6-amino-2-thiouracil monohydrate, 6-amino-2-thioxo-2,3-dihydropyrimidin-4 1h-one hydrate, 2-pyrimidinethiol, 4-amino-6-hydroxy-, monohydrate, 4-amino-6-hydroxy-2-mercaptopyrimidine hydrate, 6-amino-2-sulfanylpyrimidin-4-ol hydrate, 6-amino-2-mercaptopyrimidin-4-ol hydrate, 4 1h-pyrimidinone, 6-amino-2,3-dihydro-2-thioxo-, monohydrate, 6-amino-2-mercapto-4-pyrimidinol monohydrate, 6-amino-2-sulfanylidene-1h-pyrimidin-4-one hydrate |
PubChem CID | 2723599 |
IUPAC Name | 6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate |
SMILES | C1=C(NC(=S)NC1=O)N.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0199222
|
Thermo Scientific Chemicals
L0199222 |
100 g |
Each for $129.90
|
|
|||||
Description
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water and DMSO.
Notes
Store at 4°C. Incompatible with oxidizing agents. Avoid excessive heat and light.
Chemical Identifiers
65802-56-4 | |
161.179 | |
BWYGMIYEADAIGW-UHFFFAOYSA-N | |
2723599 | |
C1=C(NC(=S)NC1=O)N.O |
C4H7N3O2S | |
MFCD00150556 | |
4-amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 6-amino-2-thiouracil monohydrate, 6-amino-2-thioxo-2,3-dihydropyrimidin-4 1h-one hydrate, 2-pyrimidinethiol, 4-amino-6-hydroxy-, monohydrate, 4-amino-6-hydroxy-2-mercaptopyrimidine hydrate, 6-amino-2-sulfanylpyrimidin-4-ol hydrate, 6-amino-2-mercaptopyrimidin-4-ol hydrate, 4 1h-pyrimidinone, 6-amino-2,3-dihydro-2-thioxo-, monohydrate, 6-amino-2-mercapto-4-pyrimidinol monohydrate, 6-amino-2-sulfanylidene-1h-pyrimidin-4-one hydrate | |
6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate |
Specifications
65802-56-4 | |
C4H7N3O2S | |
100 g | |
4-amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 6-amino-2-thiouracil monohydrate, 6-amino-2-thioxo-2,3-dihydropyrimidin-4 1h-one hydrate, 2-pyrimidinethiol, 4-amino-6-hydroxy-, monohydrate, 4-amino-6-hydroxy-2-mercaptopyrimidine hydrate, 6-amino-2-sulfanylpyrimidin-4-ol hydrate, 6-amino-2-mercaptopyrimidin-4-ol hydrate, 4 1h-pyrimidinone, 6-amino-2,3-dihydro-2-thioxo-, monohydrate, 6-amino-2-mercapto-4-pyrimidinol monohydrate, 6-amino-2-sulfanylidene-1h-pyrimidin-4-one hydrate | |
BWYGMIYEADAIGW-UHFFFAOYSA-N | |
6-amino-2-sulfanylidene-1H-pyrimidin-4-one;hydrate | |
2723599 | |
≥98% |
>300°C | |
MFCD00150556 | |
5795441 | |
Soluble in water and DMSO. | |
C1=C(NC(=S)NC1=O)N.O | |
161.179 | |
161.19 (143.17 Anhydrous) | |
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate |
Safety and Handling
EINECSNumber : 213-722-8
RTECSNumber : YQ8817000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only