Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 90+%

$62.68 - $244.09

Chemical Identifiers

Chemical Name or Material 4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine
CAS 5472-41-3
Molecular Formula C5H5N5O
Formula Weight 151.13
MDL Number MFCD00005689
Beilstein 152192
Synonym 4-Amino-1H-pyrazolo[3,4-d]pyrimidin-6-ol; 4-Amino-1,7-dihydro-6H-pyrazolo[3,4-d]pyrimidin-6-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0369103

  SDS
alfa aesar™
L0369103
1g Each for $62.68

AAL0369106

  SDS
alfa aesar™
L0369106
5g Each for $244.09
Description & Specifications

Specifications

Formula Weight 151.13
MDL Number MFCD00005689
Chemical Name or Material 4-Amino-6-hydroxy-1H-pyrazolo[3,4-d]pyrimidine
Assay ≥90%
Molecular Formula C5H5N5O
CAS 5472-41-3
Synonym 4-Amino-1H-pyrazolo[3,4-d]pyrimidin-6-ol; 4-Amino-1,7-dihydro-6H-pyrazolo[3,4-d]pyrimidin-6-one
Quantity 1g
Odor Odorless
Beilstein 152192
Melting Point >300°C