Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-6-chloropyrimidine, 98%

$33.09 - $48.49

Chemical Identifiers

Chemical Name or Material 4-Amino-6-chloropyrimidine
CAS 5305-59-9
Molecular Formula C4H4ClN3
Formula Weight 129.55
MDL Number MFCD00053576
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5055103

 
alfa aesar™
H5055103
1g Each for $48.49

AAH50551MD

 
alfa aesar™
H50551MD
250mg Each for $33.09
Description & Specifications

Specifications

Formula Weight 129.55
Chemical Name or Material 4-Amino-6-chloropyrimidine
Quantity 1g
Assay 98%
Molecular Formula C4H4ClN3
MDL Number MFCD00053576
CAS 5305-59-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.