Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-cyano-2-methylpyrimidine, 95%

Chemical Identifiers

Chemical Name or Material 4-Amino-5-cyano-2-methylpyrimidine
CAS 698-29-3
Formula Weight 134.14
MDL Number MFCD00084875
UN Number UN3439
Synonym 4-Amino-2-methylpyrimidine-5-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6134403

  SDS
alfa aesar™
H6134403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61344MD

  SDS
alfa aesar™
H61344MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.14
Chemical Name or Material 4-Amino-5-cyano-2-methylpyrimidine
Color Pale yellow
Quantity 1g
Physical Form Powder
Assay 95%
MDL Number MFCD00084875
CAS 698-29-3
UN Number UN3439
Synonym 4-Amino-2-methylpyrimidine-5-carbonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Applications
4-Amino-5-cyano-2-methylpyrimidine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (6.4 g/L at 25°C). Soluble in dimethyl sulfoxide, ethanol and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.