Please login to your online account to display your discounted pricing

4-Amino-5-chloro-2-methoxybenzoic Acid 98+%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  133671000

 View more versions of this product

Catalog No. AC133671000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 201.61
Quantity 100g
Color White to slightly beige
EINECS Number 230-582-3
Melting Point 206° to 210°C
CAS Min % 98.0
CAS Max % 100.0
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Amino-5-chloro-2-methoxybenzoic acid
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C8H8ClNO3
Linear Formula NH2(Cl)C6H2(OCH3)COOH
MDL Number MFCD00038794
CAS 7206-70-4
Water 0.2% max. (K.F.)

Specifications

Chemical Name or Material 4-Amino-5-chloro-2-methoxybenzoic acid
Name Note 98+%
CAS 7206-70-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H8ClNO3
Linear Formula NH2(Cl)C6H2(OCH3)COOH
Formula Weight 201.61
MDL Number MFCD00038794
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

SDS & Safety Documents