Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-chloro-2,6-dimethylpyrimidine, 99%

$108.04 - $322.45

Chemical Identifiers

Chemical Name or Material 4-Amino-5-chloro-2,6-dimethylpyrimidine
CAS 2858-20-0
Molecular Formula C6H8ClN3
Formula Weight 157.6
MDL Number MFCD00191727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2249906

  SDS
alfa aesar™
B2249906
5g Each for $108.04

AAB2249914

  SDS
alfa aesar™
B2249914
25g Each for $322.45
Description & Specifications

Specifications

Formula Weight 157.6
Chemical Name or Material 4-Amino-5-chloro-2,6-dimethylpyrimidine
Melting Point 164° to 166°C
Quantity 5g
Assay 99%
Molecular Formula C6H8ClN3
MDL Number MFCD00191727
CAS 2858-20-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.