missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-bromo-6-chloropyrimidine, 98%

$53.66 - $774.15

Chemical Identifiers

CAS 663193-80-4
Molecular Formula C4H3BrClN3
Molecular Weight (g/mol) 208.443
MDL Number MFCD09999224
InChI Key DQSBRYJETNYGCI-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-6-chloropyrimidine, 5-bromo-6-chloro-4-pyrimidinamine, 5-bromo-6-chloro-pyrimidin-4-ylamine
PubChem CID 45789713
IUPAC Name 5-bromo-6-chloropyrimidin-4-amine
SMILES C1=NC(=C(C(=N1)Cl)Br)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64369MD View Documents Alfa Aesar™
H64369MD
250mg Each for $53.66
 
AAH6436903 View Documents Alfa Aesar™
H6436903
1g Each for $171.18
 
AAH6436906 View Documents Alfa Aesar™
H6436906
5g Each for $774.15
 
Specifications

Chemical Identifiers

663193-80-4
208.443
DQSBRYJETNYGCI-UHFFFAOYSA-N
45789713
C1=NC(=C(C(=N1)Cl)Br)N
C4H3BrClN3
MFCD09999224
4-amino-5-bromo-6-chloropyrimidine, 5-bromo-6-chloro-4-pyrimidinamine, 5-bromo-6-chloro-pyrimidin-4-ylamine
5-bromo-6-chloropyrimidin-4-amine

Specifications

Gray
663193-80-4
MFCD09999224
4-amino-5-bromo-6-chloropyrimidine, 5-bromo-6-chloro-4-pyrimidinamine, 5-bromo-6-chloro-pyrimidin-4-ylamine
C1=NC(=C(C(=N1)Cl)Br)N
208.443
208.44
98%
250mg
C4H3BrClN3
23483365
DQSBRYJETNYGCI-UHFFFAOYSA-N
5-bromo-6-chloropyrimidin-4-amine
45789713
Solid
4-Amino-5-bromo-6-chloropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning