Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-bromo-2-hydroxypyrimidine, 98%

Chemical Identifiers

Chemical Name or Material 4-Amino-5-bromo-2-hydroxypyrimidine
CAS 2240-25-7
Formula Weight 190
MDL Number MFCD00056025
Beilstein 127286
Synonym 5-Bromocytosine; 4-amino-5-bromopyrimidin-2(1H)-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6195203

 
alfa aesar™
H6195203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 240° to 243°C
Formula Weight 190
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Amino-5-bromo-2-hydroxypyrimidine
Physical Form Solid
Assay 98%
Color White to yellow
Quantity 1g
MDL Number MFCD00056025
CAS 2240-25-7
Beilstein 127286
Synonym 5-Bromocytosine; 4-amino-5-bromopyrimidin-2(1H)-one
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

Applications
4-Amino-5-bromo-2-hydroxypyrimidine can be used in the synthesis of cross-link products under anaerobic and aerobic conditions.

Solubility
Soluble in formic acid (50 mg/ml).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.