missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-5-bromo-2-chloropyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC429380010

 View more versions of this product

Catalog No. AC429380010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Amino-5-bromo-2-chloropyrimidine
96.0
Authentic
Liquid
97%
Clear brown to purple
208.45
MFCD07782040
205672-25-9
100.0
Glass bottle
≥96.0% (GC)
187° to 192°C
C4H3BrClN3
1g

Chemical Identifiers

205672-25-9
100.0
C4H3BrClN3
MFCD07782040
≥96.0% (GC)
96.0
97%
208.45
4-Amino-5-bromo-2-chloropyrimidine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.