missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-5-bromo-2-chloropyrimidine, 97%, ACROS Organics™

Chemical Identifiers

CAS 205672-25-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H3BrClN3
Formula Weight 208.45
MDL Number MFCD07782040
Chemical Name or Material 4-Amino-5-bromo-2-chloropyrimidine
Assay ≥96.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC429380010 SDS Thermo Fisher Scientific
AC429380010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC429380050 SDS Thermo Fisher Scientific
AC429380050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

205672-25-9
100.0
C4H3BrClN3
MFCD07782040
≥96.0% (GC)
96.0
97%
208.45
4-Amino-5-bromo-2-chloropyrimidine

Specifications

205672-25-9
100.0
Glass bottle
97%
Clear brown to purple
208.45
MFCD07782040
≥96.0% (GC)
96.0
Authentic
Liquid
187° to 192°C
C4H3BrClN3
1g
4-Amino-5-bromo-2-chloropyrimidine

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.