missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-bromo-2-chloropyrimidine, 95%

Manufacturer:  Alfa Aesar™ H6142903

 View more versions of this product

Catalog No. AAH6142903



Description

Description

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

4-Amino-5-bromo-2-chloropyrimidine
205672-25-9
MFCD07782040
QOWALNIZDHZTSM-UHFFFAOYSA-N
5-bromo-2-chloropyrimidin-4-amine
2763293
Solid
1g
C4H3BrClN3
4-amino-5-bromo-2-chloropyrimidine
C1=C(C(=NC(=N1)Cl)N)Br
208.443
208.44
95%

Chemical Identifiers

205672-25-9
208.443
QOWALNIZDHZTSM-UHFFFAOYSA-N
2763293
C1=C(C(=NC(=N1)Cl)N)Br
C4H3BrClN3
MFCD07782040
4-amino-5-bromo-2-chloropyrimidine
5-bromo-2-chloropyrimidin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents