missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-bromo-2-chloropyrimidine, 95%

Manufacturer:  Alfa Aesar™ H6142906

 View more versions of this product

Catalog No. AAH6142906



Description

Description

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

4-Amino-5-bromo-2-chloropyrimidine
205672-25-9
MFCD07782040
Slightly soluble in water.
C1=C(C(=NC(=N1)Cl)N)Br
208.443
208.44
95%
5g
C4H3BrClN3
4-amino-5-bromo-2-chloropyrimidine
QOWALNIZDHZTSM-UHFFFAOYSA-N
5-bromo-2-chloropyrimidin-4-amine
2763293
Solid

Chemical Identifiers

205672-25-9
208.443
QOWALNIZDHZTSM-UHFFFAOYSA-N
2763293
C1=C(C(=NC(=N1)Cl)N)Br
C4H3BrClN3
MFCD07782040
4-amino-5-bromo-2-chloropyrimidine
5-bromo-2-chloropyrimidin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents