Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-5-bromo-2-chloropyridine, 95%

$207.80 - $475.64

Chemical Identifiers

Chemical Name or Material 4-Amino-5-bromo-2-chloropyridine
CAS 857730-21-3
Molecular Formula C5H4BrClN2
Formula Weight 207.46
MDL Number MFCD09037459
Synonym 5-Bromo-2-chloro-4-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2767403

 
alfa aesar™
H2767403
1g Each for $475.64

AAH27674MD

 
alfa aesar™
H27674MD
250mg Each for $207.80
Description & Specifications

Specifications

Formula Weight 207.46
Chemical Name or Material 4-Amino-5-bromo-2-chloropyridine
Assay 95%
Melting Point 127° to 129°C
Molecular Formula C5H4BrClN2
Quantity 1g
MDL Number MFCD09037459
CAS 857730-21-3
Synonym 5-Bromo-2-chloro-4-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.