missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-5,6-dichloropyrimidine, 98%, Thermo Scientific™
$61.73 - $221.58
Chemical Identifiers
CAS | 310400-38-5 |
---|---|
Molecular Formula | C4H3Cl2N3 |
Molecular Weight (g/mol) | 163.99 |
MDL Number | MFCD09837293 |
InChI Key | ISTJUVRUUBNZNB-UHFFFAOYSA-N |
Synonym | 4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci |
PubChem CID | 24219262 |
IUPAC Name | 5,6-dichloropyrimidin-4-amine |
SMILES | NC1=C(Cl)C(Cl)=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6469103
|
Thermo Scientific Chemicals
H6469103 |
1 g |
Each for $61.73
|
|
|||||
AAH6469106
|
Thermo Scientific Chemicals
H6469106 |
5 g |
Each for $221.58
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
310400-38-5 | |
163.99 | |
ISTJUVRUUBNZNB-UHFFFAOYSA-N | |
24219262 | |
NC1=C(Cl)C(Cl)=NC=N1 |
C4H3Cl2N3 | |
MFCD09837293 | |
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci | |
5,6-dichloropyrimidin-4-amine |
Specifications
310400-38-5 | |
C4H3Cl2N3 | |
1 g | |
4-amino-5,6-dichloropyrimidine, 5,6-dichloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro, 5,6-dichloro-4-pyrimidinamine, 5,6-dichloropyrimidin-4-ylamine, 4-pyrimidinamine, 5,6-dichloro-9ci | |
ISTJUVRUUBNZNB-UHFFFAOYSA-N | |
5,6-dichloropyrimidin-4-amine | |
24219262 | |
98% | |
4-Amino-5,6-dichloropyrimidine |
White | |
MFCD09837293 | |
8678505 | |
Slightly soluble in water. | |
NC1=C(Cl)C(Cl)=NC=N1 | |
163.99 | |
163.99 | |
Solid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures