Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-(trifluoromethyl)pyridine, 97%

Chemical Identifiers

Chemical Name or Material 4-Amino-3-(trifluoromethyl)pyridine
CAS 387824-61-5
Molecular Formula C6H5F3N2
Formula Weight 162.11
MDL Number MFCD02183554
UN Number UN2811
Synonym 3-Trifluoromethyl-4-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL19549MC

  SDS
alfa aesar™
L19549MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19549ME

  SDS
alfa aesar™
L19549ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.11
Molecular Formula C6H5F3N2
Chemical Name or Material 4-Amino-3-(trifluoromethyl)pyridine
CAS 387824-61-5
MDL Number MFCD02183554
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 100mg
Assay 97%
Synonym 3-Trifluoromethyl-4-pyridinamine
Melting Point 98° to 102°C
Health Hazard 2 GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
UN Number UN2811
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.