Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-pyridinemethanol, 97%

$117.49 - $1,406.91

Chemical Identifiers

Chemical Name or Material 4-Amino-3-pyridinemethanol
CAS 138116-34-4
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD06203066
Synonym 4-Amino-3-(hydroxymethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5003306

 
alfa aesar™
H5003306
5g Each for $1,406.91

AAH5003303

 
alfa aesar™
H5003303
1g Each for $117.49
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 4-Amino-3-pyridinemethanol
Melting Point 167° to 169°C
Quantity 5g
Assay 97%
Molecular Formula C6H8N2O
MDL Number MFCD06203066
CAS 138116-34-4
Synonym 4-Amino-3-(hydroxymethyl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C

Applications
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C