missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-3-nitropyridine, 96%

Manufacturer:  Alfa Aesar™ L1972506

 View more versions of this product

Catalog No. AAL1972506


 

Specifications

Description

4-Amino-3-nitropyridine; 96%; Alfa Aesar; 5g; FW: 139.11; C5H5N3O2; MP: 201 to 205; CAS: 1681-37-4

Applications
4-Amino-3-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Description & Specifications

Specifications

4-Amino-3-nitropyridine
201° to 205°C
5g
139.11
383710
1681-37-4
96%
C5H5N3O2
MFCD00160304
3-Nitro-4-pyridinamine

Chemical Identifiers

1681-37-4
139.11
383710
4-Amino-3-nitropyridine
C5H5N3O2
MFCD00160304
3-Nitro-4-pyridinamine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger